Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- TBC1D15
- Entrez Gene ID
-
64786
- Gene Name
- TBC1 domain family member 15
- Gene Aliases
-
RAB7-GAP
- Chromosome Location
-
Chr.12: 71839707 - 71926849 on Build GRCh38
View Full Product Details