The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005055.4 | NP_005046.2 | 3 - 4 | 899 | 82 |
NM_032645.4 | NP_116034.2 | 3 - 4 | 899 | 82 | |
XM_005253042.3 | XP_005253099.1 | 3 - 4 | 857 | 82 | |
XM_005253043.3 | XP_005253100.1 | 3 - 4 | 857 | 82 | |
XM_011520252.1 | XP_011518554.1 | 3 - 4 | 857 | 82 | |
GenBank mRNA | AF449218.1 | - | 3 - 4 | 899 | 82 |
AK310002.1 | - | 3 - 4 | 857 | 82 | |
BC004196.2 | - | 3 - 4 | 864 | 82 | |
KU178300.1 | - | 3 - 4 | 685 | 82 | |
KU178301.1 | - | 3 - 4 | 685 | 82 | |
KU178302.1 | - | 3 - 4 | 685 | 82 | |
KU178303.1 | - | 3 - 4 | 685 | 82 | |
KU178304.1 | - | 3 - 4 | 685 | 82 |