The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080743.4 | NP_542781.3 | 1 - 2 | 258 | 97 |
XM_017010292.1 | XP_016865781.1 | 1 - 2 | 186 | 97 | |
GenBank mRNA | AK314008.1 | - | 1 - 2 | 181 | 97 |
BC021715.1 | - | 1 - 2 | 195 | 97 | |
BP244338.1 | - | 1 - 2 | 258 | 97 |