The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017855.3 | NP_060325.3 | 8 - 9 | 669 | 91 |
XM_005265698.3 | XP_005265755.1 | 9 - 10 | 733 | 91 | |
XM_006714245.3 | XP_006714308.1 | 8 - 9 | 662 | 91 | |
XM_017008331.1 | XP_016863820.1 | 7 - 8 | 625 | 91 | |
GenBank mRNA | AK000520.1 | - | 5 - 6 | 320 | 91 |
AK225547.1 | - | 5 - 6 | 300 | 91 | |
BC017796.1 | - | 4 - 5 | 276 | 91 | |
EF113908.1 | - | 8 - 9 | 671 | 91 |