The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017855.3 | NP_060325.3 | 10 - 11 | 887 | 152 |
XM_005265698.3 | XP_005265755.1 | 11 - 12 | 951 | 152 | |
XM_006714245.3 | XP_006714308.1 | 10 - 11 | 880 | 152 | |
XM_017008331.1 | XP_016863820.1 | 9 - 10 | 843 | 152 | |
GenBank mRNA | AK000520.1 | - | 7 - 8 | 538 | 152 |
AK098169.1 | - | 3 - 4 | 581 | 152 | |
AK225547.1 | - | 7 - 8 | 518 | 152 | |
BC017796.1 | - | 6 - 7 | 494 | 152 |