The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024852.3 | NP_079128.2 | 18 - 19 | 2814 | 96 |
NM_177422.2 | NP_803171.1 | 16 - 17 | 2556 | 96 | |
XM_005270575.3 | XP_005270632.1 | 19 - 20 | 8268 | 96 | |
XM_005270576.3 | XP_005270633.1 | 13 - 14 | 3052 | 96 | |
XM_011540879.2 | XP_011539181.1 | 18 - 19 | 3696 | 96 | |
XM_011540882.2 | XP_011539184.1 | 15 - 16 | 1911 | 96 | |
XM_017000523.1 | XP_016856012.1 | 19 - 20 | 8284 | 96 | |
XM_017000524.1 | XP_016856013.1 | 14 - 15 | 2082 | 96 | |
XM_017000525.1 | XP_016856014.1 | 15 - 16 | 1916 | 96 | |
XM_017000526.1 | XP_016856015.1 | 14 - 15 | 1875 | 96 | |
XM_017000527.1 | XP_016856016.1 | 14 - 15 | 2054 | 96 | |
XM_017000528.1 | XP_016856017.1 | 15 - 16 | 2003 | 96 | |
GenBank mRNA | AK022827.1 | - | 18 - 19 | 2569 | 96 |
AK292870.1 | - | 18 - 19 | 2780 | 96 | |
AK302289.1 | - | 11 - 12 | 1563 | 96 | |
AK303929.1 | - | 15 - 16 | 2098 | 96 | |
BC025769.1 | - | 16 - 17 | 2380 | 96 |