The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024852.3 | NP_079128.2 | 9 - 10 | 1498 | 109 |
NM_177422.2 | NP_803171.1 | 7 - 8 | 1240 | 109 | |
XM_005270575.3 | XP_005270632.1 | 10 - 11 | 6952 | 109 | |
XM_005270576.3 | XP_005270633.1 | 4 - 5 | 1736 | 109 | |
XM_011540879.2 | XP_011539181.1 | 9 - 10 | 2380 | 109 | |
XM_011540882.2 | XP_011539184.1 | 6 - 7 | 595 | 109 | |
XM_017000523.1 | XP_016856012.1 | 10 - 11 | 6968 | 109 | |
XM_017000524.1 | XP_016856013.1 | 5 - 6 | 766 | 109 | |
XM_017000525.1 | XP_016856014.1 | 6 - 7 | 600 | 109 | |
XM_017000526.1 | XP_016856015.1 | 5 - 6 | 559 | 109 | |
XM_017000527.1 | XP_016856016.1 | 5 - 6 | 738 | 109 | |
XM_017000528.1 | XP_016856017.1 | 6 - 7 | 687 | 109 | |
GenBank mRNA | AK022827.1 | - | 9 - 10 | 1253 | 109 |
AK292870.1 | - | 9 - 10 | 1464 | 109 | |
AK302289.1 | - | 2 - 3 | 247 | 109 | |
AK303929.1 | - | 6 - 7 | 782 | 109 | |
BC025769.1 | - | 7 - 8 | 1064 | 109 |