The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003095.2 | NP_003086.1 | 2 - 3 | 266 | 90 |
GenBank mRNA | AK311752.1 | - | 2 - 3 | 229 | 90 |
BC002505.2 | - | 2 - 3 | 259 | 90 | |
BC063397.1 | - | 2 - 3 | 216 | 90 | |
BC128452.1 | - | 2 - 3 | 169 | 90 | |
BC128453.1 | - | 2 - 3 | 168 | 90 | |
BM972965.1 | - | 2 - 3 | 90 | ||
BU861031.1 | - | 2 - 3 | 269 | 90 | |
CR456751.1 | - | 1 - 2 | 123 | 90 | |
X85372.1 | - | 2 - 3 | 238 | 90 |