Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMOD2
Entrez Gene ID
29767
Gene Name
tropomodulin 2
Gene Aliases
N-TMOD, NTMOD
Chromosome Location
Chr.15: 51751561 - 51816363 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014548.3 NP_055363.1 6 - 7 845 76
XM_017022089.1 XP_016877578.1 6 - 7 809 76
GenBank mRNA AF177169.1 - 6 - 7 780 76
AK292733.1 - 6 - 7 724 76
BC036184.1 - 7 - 8 891 76
BC064961.1 - 6 - 7 793 76

View Full Product Details