The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_199339.2 | NP_955371.2 | 2 - 3 | 235 | 90 |
GenBank mRNA | BC033882.1 | - | 2 - 3 | 235 | 90 |