Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LRRC9
Entrez Gene ID
341883
Gene Name
leucine rich repeat containing 9
Gene Aliases
-
Chromosome Location
Chr.14: 59919713 - 60063559 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_075071.1 8 - 9 1090 82
GenBank mRNA AK128037.1 - 8 - 9 1090 82

View Full Product Details