The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001301009.1 | NP_001287938.1 | 3 - 4 | 705 | 59 |
NM_001317843.1 | NP_001304772.1 | 3 - 4 | 705 | 59 | |
NM_182546.3 | NP_872352.2 | 3 - 4 | 705 | 59 | |
NR_133927.1 | 2 - 3 | 106 | 59 | ||
XM_006715665.3 | XP_006715728.1 | 3 - 4 | 966 | 59 | |
XM_006715666.3 | XP_006715729.1 | 3 - 4 | 966 | 59 | |
XM_017011841.1 | XP_016867330.1 | 3 - 4 | 966 | 59 | |
GenBank mRNA | BC028404.1 | - | 3 - 4 | 580 | 59 |
BI553644.1 | - | 3 - 4 | 512 | 59 | |
BI667944.1 | - | 2 - 3 | 113 | 59 | |
BI752069.1 | - | 2 - 3 | 121 | 59 | |
BI754879.1 | - | 3 - 4 | 245 | 59 | |
BI758069.1 | - | 3 - 4 | 367 | 59 | |
DN991717.1 | - | 3 - 4 | 467 | 59 |