The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001207015.1 | NP_001193944.1 | 10 - 11 | 1369 | 141 |
NM_001207016.1 | NP_001193945.1 | 8 - 9 | 1159 | 141 | |
NM_002849.3 | NP_002840.2 | 11 - 12 | 2025 | 141 | |
NM_130846.2 | NP_570897.2 | 7 - 8 | 1291 | 141 | |
NR_073474.1 | 7 - 8 | 1361 | 141 | ||
XM_011538615.2 | XP_011536917.1 | 11 - 12 | 1621 | 141 | |
GenBank mRNA | AK295951.1 | - | 10 - 11 | 1359 | 141 |
AK304457.1 | - | 8 - 9 | 1118 | 141 | |
AK312376.1 | - | 11 - 12 | 1938 | 141 | |
BC110900.1 | - | 7 - 8 | 988 | 141 | |
BC119777.1 | - | 8 - 9 | 967 | 141 | |
BC122866.1 | - | 8 - 9 | 967 | 141 | |
BX571751.1 | - | 7 - 8 | 1344 | 141 | |
CR749836.1 | - | 2 - 3 | 424 | 141 | |
D64053.1 | - | 11 - 12 | 2027 | 141 | |
U42361.1 | - | 11 - 12 | 1785 | 141 | |
U77916.1 | - | 8 - 9 | 1009 | 141 | |
U77917.1 | - | 7 - 8 | 1268 | 141 | |
X82635.1 | - | 7 - 8 | 1249 | 141 | |
Z79693.1 | - | 11 - 12 | 1696 | 141 |