The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144982.4 | NP_659419.3 | 20 - 21 | 4309 | 148 |
GenBank mRNA | AB011118.2 | - | 20 - 21 | 4159 | 148 |
AK125035.1 | - | 11 - 12 | 1763 | 148 | |
BX647709.1 | - | 20 - 21 | 4189 | 148 |