The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144982.4 | NP_659419.3 | 26 - 27 | 5410 | 149 |
GenBank mRNA | AB011118.2 | - | 26 - 27 | 5260 | 149 |
AK125035.1 | - | 17 - 18 | 2864 | 149 | |
BX647709.1 | - | 26 - 27 | 5290 | 149 |