Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WRN
Entrez Gene ID
7486
Gene Name
Werner syndrome RecQ like helicase
Gene Aliases
RECQ3, RECQL2, RECQL3
Chromosome Location
Chr.8: 31033262 - 31173761 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000553.4 NP_000544.2 11 - 12 2212 245
XM_011544639.2 XP_011542941.1 10 - 11 1613 164
GenBank mRNA AF091214.1 - 11 - 12 1655 245
AY818673.1 - 10 - 11 1424 245

View Full Product Details