The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165417.1 | NP_001158889.1 | 4 - 5 | 771 | 69 |
NM_001165418.1 | NP_001158890.1 | 3 - 4 | 651 | 69 | |
NM_003562.4 | NP_003553.2 | 4 - 5 | 804 | 69 | |
GenBank mRNA | AF070548.1 | - | 4 - 5 | 658 | 69 |
AK290376.1 | - | 4 - 5 | 555 | 69 | |
AK313010.1 | - | 4 - 5 | 660 | 69 | |
BC006508.2 | - | 4 - 5 | 660 | 69 | |
BC006519.2 | - | 4 - 5 | 660 | 69 | |
BC016294.2 | - | 4 - 5 | 579 | 69 | |
BC017170.2 | - | 4 - 5 | 660 | 69 | |
BI752999.1 | - | 3 - 4 | 426 | 69 | |
BM047391.1 | - | 4 - 5 | 519 | 69 | |
CR456834.1 | - | 4 - 5 | 547 | 69 |