The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004331.2 | NP_004322.1 | 4 - 5 | 586 | 99 |
XM_005273617.3 | XP_005273674.1 | 4 - 5 | 490 | 99 | |
GenBank mRNA | AB004788.1 | - | 4 - 5 | 507 | 99 |
AF060922.1 | - | 4 - 5 | 540 | 99 | |
AF067396.1 | - | 4 - 5 | 544 | 99 | |
AF079221.1 | - | 4 - 5 | 544 | 99 | |
AF452712.2 | - | 4 - 5 | 559 | 99 | |
AF536326.1 | - | 4 - 5 | 522 | 99 | |
AK290213.1 | - | 4 - 5 | 502 | 99 | |
AK300286.1 | - | 4 - 5 | 511 | 99 | |
AK307195.1 | - | 2 - 3 | 282 | 99 | |
AK315870.1 | - | 4 - 5 | 463 | 99 | |
AK316315.1 | - | 4 - 5 | 490 | 99 | |
AL132665.1 | - | 4 - 5 | 560 | 99 | |
BC001559.1 | - | 4 - 5 | 486 | 99 | |
BC009603.1 | - | 4 - 5 | 554 | 99 | |
BT019500.1 | - | 4 - 5 | 462 | 99 | |
BT019501.1 | - | 4 - 5 | 462 | 99 | |
CD558918.1 | - | 4 - 5 | 100 | ||
CR456701.1 | - | 4 - 5 | 462 | 99 |