The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127582.1 | NP_001121054.1 | 5 - 6 | 537 | 62 |
NM_001127583.1 | NP_001121055.1 | 5 - 6 | 528 | 62 | |
NM_001127584.1 | NP_001121056.1 | 5 - 6 | 525 | 62 | |
NM_001127585.1 | NP_001121057.1 | 4 - 5 | 465 | 62 | |
NM_016162.3 | NP_057246.2 | 5 - 6 | 534 | 62 | |
XM_005253698.3 | XP_005253755.1 | 6 - 7 | 785 | 62 | |
XM_011520964.2 | XP_011519266.1 | 4 - 5 | 463 | 62 | |
XM_011520965.2 | XP_011519267.1 | 5 - 6 | 484 | 62 | |
XM_017019392.1 | XP_016874881.1 | 6 - 7 | 782 | 62 | |
XM_017019393.1 | XP_016874882.1 | 6 - 7 | 773 | 62 | |
GenBank mRNA | AB197695.1 | - | 5 - 6 | 493 | 62 |
AB197696.1 | - | 5 - 6 | 487 | 62 | |
AB197697.1 | - | 5 - 6 | 484 | 62 | |
AF063594.1 | - | 5 - 6 | 512 | 62 | |
AF110645.1 | - | 5 - 6 | 513 | 62 | |
AF156552.1 | - | 5 - 6 | 513 | 62 | |
AK293169.1 | - | 4 - 5 | 450 | 62 | |
AK313621.1 | - | 5 - 6 | 506 | 62 | |
BC007781.2 | - | 5 - 6 | 499 | 62 | |
BC013038.2 | - | 5 - 6 | 479 | 62 | |
BC095434.1 | - | 5 - 6 | 498 | 62 | |
BG705367.1 | - | 5 - 6 | 525 | 62 | |
BG718170.1 | - | 5 - 6 | 545 | 62 | |
BM922497.1 | - | 6 - 7 | 763 | 62 | |
BP303219.1 | - | 5 - 6 | 534 | 62 | |
EF152349.1 | - | 4 - 5 | 424 | 62 | |
EF152350.1 | - | 4 - 5 | 329 | 62 |