The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127582.1 | NP_001121054.1 | 6 - 7 | 695 | 66 |
NM_001127583.1 | NP_001121055.1 | 6 - 7 | 686 | 66 | |
NM_001127584.1 | NP_001121056.1 | 6 - 7 | 683 | 66 | |
NM_001127585.1 | NP_001121057.1 | 5 - 6 | 623 | 66 | |
NM_016162.3 | NP_057246.2 | 6 - 7 | 692 | 66 | |
XM_005253698.3 | XP_005253755.1 | 7 - 8 | 943 | 66 | |
XM_011520964.2 | XP_011519266.1 | 5 - 6 | 621 | 66 | |
XM_011520965.2 | XP_011519267.1 | 6 - 7 | 642 | 66 | |
XM_017019392.1 | XP_016874881.1 | 7 - 8 | 940 | 66 | |
XM_017019393.1 | XP_016874882.1 | 7 - 8 | 931 | 66 | |
GenBank mRNA | AB197695.1 | - | 6 - 7 | 651 | 66 |
AB197696.1 | - | 6 - 7 | 645 | 66 | |
AB197697.1 | - | 6 - 7 | 642 | 66 | |
AF063594.1 | - | 6 - 7 | 670 | 66 | |
AF110645.1 | - | 6 - 7 | 671 | 66 | |
AF156552.1 | - | 6 - 7 | 671 | 66 | |
AK293169.1 | - | 5 - 6 | 608 | 66 | |
AK303354.1 | - | 6 - 7 | 568 | 66 | |
AK313621.1 | - | 6 - 7 | 664 | 66 | |
BC007781.2 | - | 6 - 7 | 657 | 66 | |
BC013038.2 | - | 6 - 7 | 637 | 66 | |
BC095434.1 | - | 6 - 7 | 656 | 66 | |
EF152349.1 | - | 5 - 6 | 582 | 66 | |
EF152350.1 | - | 5 - 6 | 487 | 66 | |
EF152352.1 | - | 6 - 7 | 545 | 66 |