The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001099737.2 | NP_001093207.1 | 9 - 10 | 1032 | 80 |
NM_032332.3 | NP_115708.1 | 7 - 8 | 775 | 80 | |
NR_029375.1 | 9 - 10 | 1015 | 80 | ||
GenBank mRNA | AK074525.1 | - | 9 - 10 | 1005 | 80 |
AK223195.1 | - | 7 - 8 | 725 | 80 | |
AK314164.1 | - | 9 - 10 | 1009 | 80 | |
BC005870.2 | - | 7 - 8 | 708 | 80 |