Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OR2L13
Entrez Gene ID
284521
Gene Name
olfactory receptor family 2 subfamily L member 13
Gene Aliases
OR2L14
Chromosome Location
Chr.1: 247937029 - 248100922 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175911.3 NP_787107.1 1 - 2 357 119
GenBank mRNA BC028158.1 - 1 - 2 195 119

View Full Product Details