Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC45A4
Entrez Gene ID
57210
Gene Name
solute carrier family 45 member 4
Gene Aliases
-
Chromosome Location
Chr.8: 141207166 - 141308321 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080431.2 NP_001073900.1 2 - 3 589 94
NM_001286646.1 NP_001273575.1 2 - 3 834 94
NM_001286648.1 NP_001273577.1 2 - 3 589 94
XM_011517169.2 XP_011515471.1 3 - 4 875 94
XM_011517170.2 XP_011515472.1 4 - 5 1680 94
XM_011517171.2 XP_011515473.1 3 - 4 930 94
XM_011517172.2 XP_011515474.1 3 - 4 1419 94
XM_011517173.2 XP_011515475.1 3 - 4 875 94
XM_011517174.2 XP_011515476.1 2 - 3 589 94
XM_011517177.2 XP_011515479.1 2 - 3 1127 94
XM_011517178.2 XP_011515480.1 2 - 3 3304 94
XM_011517179.2 XP_011515481.1 2 - 3 419 94
XM_017013648.1 XP_016869137.1 3 - 4 1954 94
XM_017013649.1 XP_016869138.1 3 - 4 859 94
XM_017013650.1 XP_016869139.1 2 - 3 1321 94
GenBank mRNA AK128212.1 - 2 - 3 589 94
BC090891.1 - 2 - 3 350 94
BC157840.1 - 2 - 3 331 94
BC171836.1 - 2 - 3 331 94
CX166285.1 - 2 - 3 403 94

View Full Product Details