Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TNNT1
Entrez Gene ID
7138
Gene Name
troponin T1, slow skeletal type
Gene Aliases
ANM, NEM5, STNT, TNT, TNTS
Chromosome Location
Chr.19: 55132698 - 55149354 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126132.2 NP_001119604.1 8 - 9 516 61
NM_001126133.2 NP_001119605.1 7 - 8 483 61
NM_001291774.1 NP_001278703.1 8 - 9 578 61
NM_003283.5 NP_003274.3 8 - 9 516 61
XM_011527246.2 XP_011525548.1 7 - 8 393 61
XM_017027186.1 XP_016882675.1 9 - 10 589 61
XM_017027187.1 XP_016882676.1 8 - 9 577 61
GenBank mRNA AK291352.1 - 8 - 9 368 61
AK308199.1 - 7 - 8 335 61
AY762903.1 - 7 - 8 311 61
AY762904.1 - 6 - 7 278 61
BC007074.1 - 7 - 8 356 61
BC010963.1 - 8 - 9 369 61
BC022086.2 - 8 - 9 417 61
BC034143.1 - 8 - 9 368 61
BC107798.1 - 7 - 8 338 61
BT019630.1 - 6 - 7 278 61
DC385627.1 - 8 - 9 516 61
DC385695.1 - 8 - 9 401 61
M19308.1 - 7 - 8 336 61
M19309.1 - 8 - 9 368 61
S51938.1 - 6 - 7 278 61
S69208.1 - 8 - 9 352 61
S69209.1 - 7 - 8 319 61

View Full Product Details