Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNNT1
Entrez Gene ID
7138
Gene Name
troponin T1, slow skeletal type
Gene Aliases
ANM, NEM5, STNT, TNT, TNTS
Chromosome Location
Chr.19: 55132698 - 55149354 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126132.2 NP_001119604.1 9 - 10 588 65
NM_001126133.2 NP_001119605.1 8 - 9 555 65
NM_001291774.1 NP_001278703.1 9 - 10 650 65
NM_003283.5 NP_003274.3 9 - 10 588 65
XM_011527246.2 XP_011525548.1 8 - 9 465 65
XM_017027186.1 XP_016882675.1 10 - 11 661 65
XM_017027187.1 XP_016882676.1 9 - 10 649 65
GenBank mRNA AK291352.1 - 9 - 10 440 65
AK308199.1 - 8 - 9 407 65
AY762903.1 - 8 - 9 383 65
AY762904.1 - 7 - 8 350 65
BC007074.1 - 8 - 9 428 65
BC010963.1 - 9 - 10 441 65
BC022086.2 - 9 - 10 489 65
BC034143.1 - 9 - 10 440 65
BC107798.1 - 8 - 9 410 65
BT019630.1 - 7 - 8 350 65
DC385695.1 - 9 - 10 473 65
M19308.1 - 8 - 9 408 65
M19309.1 - 9 - 10 440 65
S69208.1 - 9 - 10 424 65
S69209.1 - 8 - 9 391 65

View Full Product Details