Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANK3
Entrez Gene ID
288
Gene Name
ankyrin 3, node of Ranvier (ankyrin G)
Gene Aliases
ANKYRIN-G, MRT37
Chromosome Location
Chr.10: 60026298 - 60733526 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001149.3 NP_001140.2 19 - 20 3590 90
NM_001204403.1 NP_001191332.1 42 - 43 5728 90
NM_001204404.1 NP_001191333.1 42 - 43 5971 90
NM_001320874.1 NP_001307803.1 41 - 42 5985 90
NM_020987.4 NP_066267.2 42 - 43 13515 90
XM_005269715.3 XP_005269772.1 42 - 43 5872 90
XM_006717802.3 XP_006717865.1 44 - 45 5911 90
XM_011539708.2 XP_011538010.1 45 - 46 8494 90
XM_011539709.2 XP_011538011.1 44 - 45 8185 90
XM_017016110.1 XP_016871599.1 45 - 46 13729 90
XM_017016111.1 XP_016871600.1 44 - 45 13717 90
XM_017016112.1 XP_016871601.1 45 - 46 13714 90
XM_017016113.1 XP_016871602.1 44 - 45 13702 90
XM_017016114.1 XP_016871603.1 45 - 46 13828 90
XM_017016115.1 XP_016871604.1 45 - 46 13558 90
XM_017016116.1 XP_016871605.1 44 - 45 13420 90
XM_017016118.1 XP_016871607.1 45 - 46 12388 90
XM_017016119.1 XP_016871608.1 44 - 45 12079 90
XM_017016121.1 XP_016871610.1 43 - 44 12052 90
XM_017016122.1 XP_016871611.1 42 - 43 8401 90
XM_017016124.1 XP_016871613.1 43 - 44 8158 90
XM_017016126.1 XP_016871615.1 42 - 43 8053 90
XM_017016127.1 XP_016871616.1 41 - 42 8026 90
XM_017016128.1 XP_016871617.1 43 - 44 5884 90
XM_017016131.1 XP_016871620.1 41 - 42 5803 90
XM_017016132.1 XP_016871621.1 41 - 42 5782 90
XM_017016133.1 XP_016871622.1 41 - 42 5917 90
XM_017016134.1 XP_016871623.1 41 - 42 5752 90
XM_017016135.1 XP_016871624.1 41 - 42 5881 90
XM_017016136.1 XP_016871625.1 43 - 44 5602 90
XM_017016137.1 XP_016871626.1 42 - 43 5575 90
XM_017016139.1 XP_016871628.1 40 - 41 5509 90
XM_017016141.1 XP_016871630.1 41 - 42 5470 90
XM_017016142.1 XP_016871631.1 40 - 41 5458 90
XM_017016143.1 XP_016871632.1 40 - 41 5443 90
XM_017016145.1 XP_016871634.1 39 - 40 5431 90
XM_017016147.1 XP_016871636.1 40 - 41 5422 90
XM_017016148.1 XP_016871637.1 40 - 41 5325 90
XM_017016150.1 XP_016871639.1 42 - 43 13354 90
GenBank mRNA AK092361.1 - 9 - 10 1315 90
AK124230.1 - 3 - 4 2241 90
AK128784.1 - 1 - 2 232 90
AK295661.1 - 42 - 43 5678 90
AL136710.1 - 19 - 20 3248 90
BX537917.1 - 42 - 43 5599 90
BX648574.1 - 42 - 43 5693 90
U13616.1 - 42 - 43 13261 90

View Full Product Details