The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022153.1 | NP_071436.1 | 3 - 4 | 715 | 92 |
GenBank mRNA | AF289594.1 | - | - | 1151 | 92 |
AK024449.1 | - | 3 - 4 | 633 | 92 | |
AY265805.1 | - | 3 - 4 | 630 | 92 | |
AY358379.1 | - | 3 - 4 | 715 | 92 | |
BC020568.1 | - | 3 - 4 | 587 | 92 | |
BC111048.1 | - | 3 - 4 | 563 | 92 | |
EF506490.1 | - | 3 - 4 | 627 | 92 |