Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EXOSC4
Entrez Gene ID
54512
Gene Name
exosome component 4
Gene Aliases
RRP41, RRP41A, Rrp41p, SKI6, Ski6p, hRrp41p, p12A
Chromosome Location
Chr.8: 144064092 - 144080648 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019037.2 NP_061910.1 1 - 2 278 106
GenBank mRNA AF281133.1 - 1 - 2 271 106
AK000598.1 - 1 - 2 195 106
BC002777.2 - 1 - 2 192 106
BM839994.1 - 1 - 2 278 106

View Full Product Details