The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002308.3 | NP_002299.2 | 8 - 9 | 776 | 62 |
NM_009587.2 | NP_033665.1 | 9 - 10 | 872 | 62 | |
NR_024043.1 | 8 - 9 | 705 | 62 | ||
XM_006721892.2 | XP_006721955.1 | 7 - 8 | 703 | 62 | |
XM_017024623.1 | XP_016880112.1 | 8 - 9 | 828 | 62 | |
GenBank mRNA | AB003517.1 | - | 8 - 9 | 683 | 62 |
AB005894.1 | - | 8 - 9 | 717 | 62 | |
AB006782.1 | - | 9 - 10 | 835 | 62 | |
AB008492.1 | - | 9 - 10 | 779 | 62 | |
AK126017.1 | - | 8 - 9 | 2126 | 62 | |
AK223232.1 | - | 8 - 9 | 740 | 62 | |
AK296031.1 | - | 8 - 9 | 647 | 62 | |
BC105942.1 | - | 8 - 9 | 700 | 62 | |
BC105944.1 | - | 8 - 9 | 700 | 62 | |
BC110340.1 | - | 8 - 9 | 717 | 62 | |
Z49107.1 | - | 8 - 9 | 745 | 62 |