The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002496.3 | NP_002487.1 | 6 - 7 | 622 | 80 |
XM_005274013.1 | XP_005274070.1 | 6 - 7 | 719 | 80 | |
XM_005274014.2 | XP_005274071.1 | 6 - 7 | 746 | 80 | |
XM_011545053.2 | XP_011543355.1 | 6 - 7 | 657 | 80 | |
GenBank mRNA | AK223114.1 | - | 6 - 7 | 612 | 80 |
AK314546.1 | - | 6 - 7 | 547 | 80 | |
BC119753.1 | - | 2 - 3 | 179 | 80 | |
BC119754.2 | - | 6 - 7 | 551 | 80 | |
U65579.1 | - | 6 - 7 | 545 | 80 |