Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PNKD
Entrez Gene ID
25953
Gene Name
paroxysmal nonkinesigenic dyskinesia
Gene Aliases
BRP17, DYT8, FKSG19, FPD1, KIPP1184, MR-1, MR1, PDC, PKND1, TAHCCP2
Chromosome Location
Chr.2: 218270392 - 218346793 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077399.2 NP_001070867.1 1 - 2 211 106
NM_015488.4 NP_056303.3 1 - 2 211 106
XM_017003771.1 XP_016859260.1 1 - 2 118 106
GenBank mRNA AF318057.1 - 1 - 2 211 106
AF417001.1 - 1 - 2 211 106
AK289867.1 - 1 - 2 119 106
AL080092.1 - 1 - 2 93 106
AY039043.1 - - 99 106
BC021118.1 - 1 - 2 98 106

View Full Product Details