Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RFTN2
Entrez Gene ID
130132
Gene Name
raftlin family member 2
Gene Aliases
C2orf11, Raftlin-2
Chromosome Location
Chr.2: 197568224 - 197676050 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_144629.2 NP_653230.2 5 - 6 1322 148
XM_011510595.2 XP_011508897.1 5 - 6 1512 148
XM_011510596.2 XP_011508898.1 5 - 6 1512 148
XM_011510597.2 XP_011508899.1 5 - 6 1512 148
XM_011510598.2 XP_011508900.1 5 - 6 1461 148
XM_017003332.1 XP_016858821.1 5 - 6 1461 148
GenBank mRNA AK055136.1 - 5 - 6 1322 148
BC093966.1 - 5 - 6 969 148
BC111069.1 - 5 - 6 1454 148
BC113361.1 - 5 - 6 969 148
BX647618.1 - 2 - 3 349 148

View Full Product Details