Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LARS
Entrez Gene ID
51520
Gene Name
leucyl-tRNA synthetase
Gene Aliases
HSPC192, ILFS1, LARS1, LEURS, LEUS, LFIS, LRS, PIG44, RNTLS, hr025Cl
Chromosome Location
Chr.5: 146113026 - 146182787 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001317964.1 NP_001304893.1 27 - 28 3146 133
NM_001317965.1 NP_001304894.1 27 - 28 3165 133
NM_016460.3 NP_057544.2 27 - 28 3203 133
NM_020117.10 NP_064502.9 28 - 29 3284 133
XM_011537656.2 XP_011535958.1 28 - 29 3150 133
GenBank mRNA AB037773.1 - 28 - 29 3157 133
AK000277.1 - 14 - 15 1508 133
AK001457.1 - 9 - 10 1043 133
AK001682.1 - 6 - 7 790 133
AK021413.1 - 7 - 8 831 133
AK023777.1 - 21 - 22 2268 133
AK025441.1 - 28 - 29 3120 133
AK127009.1 - 4 - 5 2969 133
AK225372.1 - 27 - 28 2982 133
AK293750.1 - 27 - 28 3033 133
AK295874.1 - 27 - 28 3001 133
AK304134.1 - 27 - 28 2962 133
AK315178.1 - 28 - 29 3133 133
AL390155.1 - 6 - 7 723 133
AY513284.1 - 9 - 10 971 133
AY926480.1 - 9 - 10 971 133
BC131798.1 - 14 - 15 1463 133
BC150213.1 - 28 - 29 3135 133
BC151214.1 - 28 - 29 3085 133
BC152422.1 - 28 - 29 3157 133
D84223.1 - 28 - 29 3062 133

View Full Product Details