Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPATA24
Entrez Gene ID
202051
Gene Name
spermatogenesis associated 24
Gene Aliases
CCDC161, T6441
Chromosome Location
Chr.5: 139395269 - 139404204 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_194296.1 NP_919272.1 5 - 6 92
GenBank mRNA AA399058.1 - 1 - 2 165 92
BC040037.1 - 5 - 6 92

View Full Product Details