Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RB1CC1
Entrez Gene ID
9821
Gene Name
RB1 inducible coiled-coil 1
Gene Aliases
ATG17, CC1, FIP200, PPP1R131
Chromosome Location
Chr.8: 52622458 - 52714466 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083617.1 NP_001077086.1 8 - 9 1732 112
NM_014781.4 NP_055596.3 8 - 9 1732 112
XM_011517643.2 XP_011515945.1 9 - 10 1921 112
XM_011517644.2 XP_011515946.1 8 - 9 1732 112
XM_011517645.1 XP_011515947.1 8 - 9 1732 112
XM_011517646.2 XP_011515948.1 7 - 8 1571 112
XM_011517647.2 XP_011515949.1 8 - 9 1732 112
XM_011517649.2 XP_011515951.1 8 - 9 1732 112
XM_017014103.1 XP_016869592.1 8 - 9 1732 112
XM_017014104.1 XP_016869593.1 9 - 10 1952 112
XM_017014105.1 XP_016869594.1 8 - 9 1732 112
XM_017014106.1 XP_016869595.1 8 - 9 1732 112
XM_017014107.1 XP_016869596.1 9 - 10 1926 112
XM_017014108.1 XP_016869597.1 8 - 9 1732 112
XM_017014109.1 XP_016869598.1 7 - 8 1584 112
XM_017014110.1 XP_016869599.1 8 - 9 1732 112
XM_017014111.1 XP_016869600.1 8 - 9 1732 112
XM_017014112.1 XP_016869601.1 8 - 9 1732 112
GenBank mRNA AB059622.1 - 8 - 9 1698 112
AK291699.1 - 8 - 9 1689 112
BC017556.1 - 7 - 8 1296 112
D86958.1 - 8 - 9 1689 112

View Full Product Details