Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RB1CC1
- Entrez Gene ID
-
9821
- Gene Name
- RB1 inducible coiled-coil 1
- Gene Aliases
-
ATG17, CC1, FIP200, PPP1R131
- Chromosome Location
-
Chr.8: 52622458 - 52714466 on Build GRCh38
View Full Product Details