Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPSF3
Entrez Gene ID
51692
Gene Name
cleavage and polyadenylation specific factor 3
Gene Aliases
CPSF-73, CPSF73
Chromosome Location
Chr.2: 9423537 - 9473110 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321833.1 NP_001308762.1 15 - 16 2252 138
NM_001321834.1 NP_001308763.1 15 - 16 3160 138
NM_001321835.1 NP_001308764.1 15 - 16 1895 138
NM_001321836.1 NP_001308765.1 16 - 17 2342 138
NM_016207.3 NP_057291.1 15 - 16 2023 138
GenBank mRNA AF171877.1 - 15 - 16 1801 138
AK025440.1 - 13 - 14 1682 138
AK223487.1 - 15 - 16 1821 138
AK298918.1 - 13 - 14 1616 138
BC011654.2 - 15 - 16 1896 138
BC014106.1 - 15 - 16 2813 138
BC020211.1 - 15 - 16 1821 138
EF036508.1 - 2 - 3 183 138

View Full Product Details