Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPSF3
Entrez Gene ID
51692
Gene Name
cleavage and polyadenylation specific factor 3
Gene Aliases
CPSF-73, CPSF73
Chromosome Location
Chr.2: 9423537 - 9473110 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001321833.1 NP_001308762.1 7 - 8 1222 92
NM_001321834.1 NP_001308763.1 7 - 8 2130 92
NM_001321835.1 NP_001308764.1 7 - 8 865 92
NM_001321836.1 NP_001308765.1 8 - 9 1312 92
NM_016207.3 NP_057291.1 7 - 8 993 92
GenBank mRNA AK025440.1 - 5 - 6 652 92
AK298918.1 - 5 - 6 586 92
BC011654.2 - 7 - 8 866 92
BC014106.1 - 7 - 8 1783 92
BC020211.1 - 7 - 8 791 92
BC030988.1 - 7 - 8 879 92

View Full Product Details