The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144061.1 | NP_001137533.1 | 20 - 21 | 2896 | 158 |
NM_001144062.1 | NP_001137534.1 | 20 - 21 | 2835 | 158 | |
NM_016451.4 | NP_057535.1 | 20 - 21 | 2957 | 158 | |
GenBank mRNA | AF111807.1 | - | 20 - 21 | 2763 | 158 |
AL136593.1 | - | 20 - 21 | 2828 | 158 | |
BC037280.2 | - | 20 - 21 | 2951 | 158 |