The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001144061.1 | NP_001137533.1 | 9 - 10 | 1308 | 143 |
NM_001144062.1 | NP_001137534.1 | 9 - 10 | 1247 | 143 | |
NM_016451.4 | NP_057535.1 | 9 - 10 | 1369 | 143 | |
GenBank mRNA | AF084457.1 | - | 9 - 10 | 1207 | 143 |
AF111807.1 | - | 9 - 10 | 1175 | 143 | |
AL136593.1 | - | 9 - 10 | 1240 | 143 | |
BC037280.2 | - | 9 - 10 | 1363 | 143 |