Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTI2
Entrez Gene ID
80185
Gene Name
TELO2 interacting protein 2
Gene Aliases
C8orf41, MRT39
Chromosome Location
Chr.8: 33498509 - 33513601 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001102401.2 NP_001095871.1 6 - 7 1463 83
NM_001265581.1 NP_001252510.1 6 - 7 1585 83
NM_025115.3 NP_079391.2 5 - 6 1832 83
XM_011544665.2 XP_011542967.1 5 - 6 1786 83
GenBank mRNA AK026916.1 - 5 - 6 1685 83
BC007387.2 - 6 - 7 1388 83
BC066935.1 - 6 - 7 1416 83
BX648410.1 - 7 - 8 1475 83

View Full Product Details