Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NPHP3-AS1
- Entrez Gene ID
-
348808
- Gene Name
- NPHP3 antisense RNA 1
- Gene Aliases
-
NCRNA00119
- Chromosome Location
-
Chr.3: 132722342 - 132874206 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC036463.1 |
- |
1 - 1 |
57 |
82 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BG721288.1 |
- |
1 |
57 |
|
Chr. 3 - 132721750 - 132722380 [+] on Build GRCh38 |
82 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
NPHP3-ACAD11 readthrough (NMD candidate) |
NPHP3-ACAD11 |
|
|
Anti-sense |
Chr. - 132558138 - 132722459 on Build GRCh38 |
Downstream |
transmembrane protein 108 |
TMEM108 |
CT124 |
315.9kb
|
Sense |
Chr. - 133038288 - 133397775 on Build GRCh38 |
View Full Product Details