Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAAP100
Entrez Gene ID
80233
Gene Name
Fanconi anemia core complex associated protein 100
Gene Aliases
C17orf70
Chromosome Location
Chr.17: 81539885 - 81552403 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025161.5 NP_079437.5 5 - 6 2248 130
NR_033338.1 5 - 6 2444 130
XM_006722112.3 XP_006722175.1 5 - 6 2394 130
XM_017025162.1 XP_016880651.1 5 - 6 2269 130
XM_017025163.1 XP_016880652.1 5 - 6 2165 130
GenBank mRNA AK024644.1 - 4 - 5 675 130
AK025828.1 - 1 - 2 509 130
AK054713.1 - 1 - 2 1592 130
AK225370.1 - 1 - 2 509 130
AL834374.1 - 1 - 2 733 130
BC008883.2 - 5 - 6 2417 130
BC021968.2 - 1 - 2 320 130
BC117139.1 - 3 - 4 1768 130
BC117141.1 - 3 - 4 1768 130
BC143861.1 - 3 - 4 1768 130
DQ989324.1 - 5 - 6 2230 130

View Full Product Details