Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC130
Entrez Gene ID
81576
Gene Name
coiled-coil domain containing 130
Gene Aliases
-
Chromosome Location
Chr.19: 13731760 - 13763298 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320561.1 NP_001307490.1 4 - 5 689 105
NM_001320564.1 NP_001307493.1 4 - 5 677 105
NM_001320565.1 NP_001307494.1 4 - 5 559 105
NM_001320566.1 NP_001307495.1 4 - 5 677 105
NM_001320567.1 NP_001307496.1 4 - 5 540 105
NM_001320568.1 NP_001307497.1 3 - 4 349 105
NM_030818.3 NP_110445.1 3 - 4 349 105
XM_005260086.4 XP_005260143.1 4 - 5 539 105
XM_011528326.1 XP_011526628.1 3 - 4 428 105
GenBank mRNA AF250306.1 - 3 - 4 277 105
AK098262.1 - 3 - 4 306 105
AK293927.1 - 4 - 5 540 105
BC002905.2 - 3 - 4 273 105
BM547551.1 - 3 - 4 347 105
BX394050.2 - 4 - 5 670 105
BX422326.2 - 3 - 4 348 105
CR999732.1 - 2 - 3 274 105

View Full Product Details