Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC130
Entrez Gene ID
81576
Gene Name
coiled-coil domain containing 130
Gene Aliases
-
Chromosome Location
Chr.19: 13731760 - 13763298 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320561.1 NP_001307490.1 5 - 6 780 79
NM_001320564.1 NP_001307493.1 5 - 6 768 79
NM_001320565.1 NP_001307494.1 5 - 6 650 79
NM_001320566.1 NP_001307495.1 5 - 6 768 79
NM_001320567.1 NP_001307496.1 5 - 6 631 79
NM_001320568.1 NP_001307497.1 4 - 5 440 79
NM_001320569.1 NP_001307498.1 3 - 4 236 79
NM_030818.3 NP_110445.1 4 - 5 440 79
XM_005260086.4 XP_005260143.1 5 - 6 630 79
XM_011528326.1 XP_011526628.1 4 - 5 519 79
GenBank mRNA AF250306.1 - 4 - 5 368 79
AK098262.1 - 4 - 5 397 79
AK293927.1 - 5 - 6 631 79
AL560878.3 - 5 - 6 651 79
BC002905.2 - 4 - 5 364 79
BI913255.1 - 3 - 4 201 79
BM547551.1 - 4 - 5 438 79
BX394050.2 - 5 - 6 761 79
BX422326.2 - 4 - 5 439 79
CR999732.1 - 3 - 4 365 79

View Full Product Details