Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCDC130
Entrez Gene ID
81576
Gene Name
coiled-coil domain containing 130
Gene Aliases
-
Chromosome Location
Chr.19: 13731760 - 13763298 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320561.1 NP_001307490.1 8 - 9 1032 88
NM_001320564.1 NP_001307493.1 8 - 9 1020 88
NM_001320565.1 NP_001307494.1 8 - 9 902 88
NM_001320569.1 NP_001307498.1 6 - 7 488 88
NM_030818.3 NP_110445.1 7 - 8 692 88
XM_005260086.4 XP_005260143.1 8 - 9 882 88
XM_011528326.1 XP_011526628.1 7 - 8 771 88
GenBank mRNA AF250306.1 - 7 - 8 620 88
AK098262.1 - 7 - 8 649 88
BC002905.2 - 7 - 8 616 88
BI913255.1 - 6 - 7 453 88
BM547551.1 - 7 - 8 690 88

View Full Product Details