The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003954.4 | NP_003945.2 | 10 - 11 | 1943 | 81 |
XM_011525441.2 | XP_011523743.1 | 11 - 12 | 2080 | 81 | |
GenBank mRNA | AK131438.1 | - | 2 - 3 | 625 | 81 |
AK290203.1 | - | 10 - 11 | 1919 | 81 | |
AK313085.1 | - | 10 - 11 | 1919 | 81 | |
BC035576.1 | - | 10 - 11 | 1888 | 81 | |
Y10256.1 | - | 9 - 10 | 2050 | 81 |