The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003954.4 | NP_003945.2 | 1 - 2 | 104 | 79 |
GenBank mRNA | AK290203.1 | - | 1 - 2 | 80 | 79 |
AK313085.1 | - | 1 - 2 | 80 | 79 | |
BC035576.1 | - | 1 - 2 | 49 | 79 | |
DA856801.1 | - | 1 - 2 | 88 | 79 | |
HY096069.1 | - | 1 - 2 | 107 | 79 |