The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003954.4 | NP_003945.2 | 3 - 4 | 452 | 78 |
XM_011525441.2 | XP_011523743.1 | 4 - 5 | 589 | 78 | |
GenBank mRNA | AK290203.1 | - | 3 - 4 | 428 | 78 |
AK313085.1 | - | 3 - 4 | 428 | 78 | |
BC035576.1 | - | 3 - 4 | 397 | 78 | |
DA856801.1 | - | 3 - 4 | 436 | 78 | |
Y10256.1 | - | 2 - 3 | 559 | 78 |