Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TMA16
Entrez Gene ID
55319
Gene Name
translation machinery associated 16 homolog
Gene Aliases
C4orf43
Chromosome Location
Chr.4: 163494521 - 163520539 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018352.2 NP_060822.2 2 - 3 404 85
GenBank mRNA AK002046.1 - 2 - 3 235 85
AK298235.1 - 2 - 3 290 86
BC011842.2 - 2 - 3 221 85
BU159489.1 - 2 - 3 404 85

View Full Product Details